Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000137.2 | NP_000128.1 | 11 | 1092 | ||
GenBank mRNA | AK313951.1 | - | 11 | 1087 | ||
BC002527.1 | - | 11 | 930 | 3139988 | ||
BT007160.1 | - | 11 | 928 | |||
BX537608.1 | - | 10 | 2957 | |||
M55150.1 | - | 11 | 984 | |||
S63548.1 | - | 1 | 26 | |||
S63549.1 | - | 1 | 26 | |||
X51728.1 | - | 10 | 855 |