Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040273.2 | NP_001035363.1 | 2 | 1337 | ||
NM_173555.3 | NP_775826.2 | 4 | 1654 | |||
NR_073580.1 | 2 | 105 | ||||
NR_073581.1 | 4 | 542 | ||||
NR_073582.1 | 4 | 422 | ||||
NR_073591.1 | 4 | 333 | ||||
NR_073592.1 | 2 | 225 | ||||
NR_073593.1 | 4 | 325 | ||||
NR_073594.1 | 4 | 445 | ||||
NR_073595.1 | 4 | 578 | ||||
GenBank mRNA | AK074968.1 | - | 4 | 480 | ||
AK097697.1 | - | 3 | 1197 | |||
BC016840.1 | - | 4 | 1108 | |||
BC030242.1 | - | 2 | 1253 | 5176098 | ||
BC047424.1 | - | 2 | 105 | |||
BC111501.1 | - | 4 | 1593 | 5732356 | ||
BI829894.1 | - | 4 | 330 | |||
CD671353.1 | - | 4 | 264 | |||
CV805418.1 | - | 4 | 578 | |||
DA334145.1 | - | 4 | 381 | |||
DA434127.1 | - | 4 | 371 | |||
HY013216.1 | - | 2 | 196 |