Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145290.1 | NP_001138762.1 | 9 | 1059 | ||
NM_198277.2 | NP_938018.1 | 9 | 1059 | |||
GenBank mRNA | AK074100.1 | - | 9 | 841 | ||
AK074207.1 | - | 9 | 1059 | |||
AK290314.1 | - | 9 | 871 | |||
BC051314.1 | - | 9 | 840 | 6207707 |