Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF438 SIRNA Assays ›
Gene Symbol:
ZNF438
Gene Name:
zinc finger protein 438
Gene Aliases:
bA330O11.1
Chromosome Location:
Chr.10: 30844634 - 31032406 on Build GRCh38
UniGene ID:
Hs.660642
Species:
Human
siRNA ID
HSS176781
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF438
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143766.1
NP_001137238.1
7
1327
NM_001143767.1
NP_001137239.1
5
1140
NM_001143768.1
NP_001137240.1
6
1198
NM_001143769.1
NP_001137241.1
7
1452
NM_001143770.1
NP_001137242.1
8
1378
NM_001143771.1
NP_001137243.1
7
1349
NM_182755.2
NP_877432.1
7
1279
XM_006717393.3
XP_006717456.1
5
1094
XM_006717396.3
XP_006717459.1
4
1028
XM_006717398.3
XP_006717461.1
8
1319
XM_006717399.3
XP_006717462.1
7
1252
XM_011519371.2
XP_011517673.1
6
3302
XM_011519372.2
XP_011517674.1
6
1329
XM_011519373.2
XP_011517675.1
5
3244
XM_011519374.2
XP_011517676.1
3
1071
XM_011519376.2
XP_011517678.1
8
1404
XM_011519377.2
XP_011517679.1
8
1400
XM_017015863.1
XP_016871352.1
3
894
XM_017015864.1
XP_016871353.1
8
3538
XM_017015865.1
XP_016871354.1
7
3479
XM_017015866.1
XP_016871355.1
7
1369
XM_017015867.1
XP_016871356.1
6
3284
XM_017015868.1
XP_016871357.1
5
1590
XM_017015869.1
XP_016871358.1
5
3225
XM_017015870.1
XP_016871359.1
6
2124
XM_017015871.1
XP_016871360.1
5
1584
XM_017015872.1
XP_016871361.1
6
3282
XM_017015873.1
XP_016871362.1
7
3459
XM_017015874.1
XP_016871363.1
5
1145
GenBank mRNA
AF428258.1
-
6
1174
AK131357.1
-
7
1452
AK292730.1
-
7
1245
AL833056.1
-
2
888
BC101622.1
-
3
904
8069128
BC104757.1
-
3
904
8143760
BC143462.1
-
4
1003
BC143464.1
-
4
1083
DQ356011.1
-
7
1273
DQ356012.1
-
7
1264
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0