Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152731.2 | NP_689944.2 | 3, 4 | 874 | ||
XM_011514346.2 | XP_011512648.1 | 3, 4 | 862 | |||
XM_017010406.1 | XP_016865895.1 | 3, 4 | 425 | |||
XM_017010407.1 | XP_016865896.1 | 3, 4 | 408 | |||
XM_017010408.1 | XP_016865897.1 | 4, 5 | 955 | |||
GenBank mRNA | AK054724.1 | - | 3, 4 | 559 | ||
AK303705.1 | - | 2, 3 | 244 | |||
DC411761.1 | - | 2, 3 | 244 |