Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040274.2 | NP_001035364.2 | 26, 27 | 2545 | ||
GenBank mRNA | AK128130.1 | - | 26, 27 | 2599 | ||
AM293674.1 | - | 26, 27 | 2264 | |||
BC012225.1 | - | 16, 17 | 1442 |