Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
siRNA ID
HSS176875
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
5
951
NM_001287743.1
NP_001274672.1
4
753
NM_001287744.1
NP_001274673.1
3
580
NM_001287745.1
NP_001274674.1
4
659
NM_001287747.1
NP_001274676.1
3
616
NM_001287748.1
NP_001274677.1
3
278
NM_001287749.1
NP_001274678.1
3
516
NM_001287750.1
NP_001274679.1
2
518
NM_001287751.1
NP_001274680.1
2
390
NM_004462.4
NP_004453.3
3
462
XM_017013217.1
XP_016868706.1
5
1243
GenBank mRNA
AK057726.1
-
3
580
AK098682.1
-
1036
AK293545.1
-
3
516
AK296043.1
-
3
334
AK297868.1
-
2
461
AK300059.1
-
2
280
AK300245.1
-
3
239
AK311246.1
-
2
229
AK311362.1
-
3
648
AK315993.1
-
4
529
AK316033.1
-
3
483
AK316182.1
-
2
478
AK316351.1
-
3
243
AK316531.1
-
3
228
AK316534.1
-
2
390
BC003573.1
-
3
309
3538137
BC009251.2
-
3
309
2961623
BC029641.1
-
3
272
4514761
BC034440.1
-
3
272
BE384903.1
-
3
616
BG752446.1
-
326
BI765996.1
-
5
555
BT006704.1
-
3
240
CR457033.1
-
3
240
DC314061.1
-
3
438
DC387127.1
-
2
512
DC387479.1
-
3
278
DC396859.1
-
3
464
L06070.1
-
3
249
L06105.1
-
3
284
S76822.1
-
3
305
X69141.1
-
3
331
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0