Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321931.1 | NP_001308860.1 | 6 | 993 | ||
NM_001321932.1 | NP_001308861.1 | 6 | 1079 | |||
NM_001321933.1 | NP_001308862.1 | 6 | 989 | |||
NM_001321934.1 | NP_001308863.1 | 5 | 752 | |||
NM_001321935.1 | NP_001308864.1 | 5 | 605 | |||
NM_001321936.1 | NP_001308865.1 | 6 | 932 | |||
NM_001321938.1 | NP_001308867.1 | 7 | 980 | |||
NM_001321939.1 | NP_001308868.1 | 4 | 987 | |||
NM_001321940.1 | NP_001308869.1 | 6 | 1335 | |||
NM_001321941.1 | NP_001308870.1 | 6 | 1167 | |||
NM_001321942.1 | NP_001308871.1 | 6 | 1174 | |||
NM_001321943.1 | NP_001308872.1 | 6 | 1211 | |||
NM_001321944.1 | NP_001308873.1 | 6 | 1427 | |||
NM_001321945.1 | NP_001308874.1 | 6 | 970 | |||
NM_001321946.1 | NP_001308875.1 | 7 | 1076 | |||
NM_001321947.1 | NP_001308876.1 | 6 | 894 | |||
NM_001321948.1 | NP_001308877.1 | 6 | 966 | |||
NM_001321949.1 | NP_001308878.1 | 6 | 1663 | |||
NM_004115.3 | NP_004106.1 | 5 | 729 | |||
NM_175929.2 | NP_787125.1 | 5 | 840 | |||
GenBank mRNA | AY188178.1 | - | 5 | 840 | ||
BC100920.1 | - | 5 | 839 | 40003901 | ||
BC100921.1 | - | 5 | 839 | 40003902 | ||
BC100922.2 | - | 5 | 839 | 40003903 | ||
U66200.1 | - | 5 | 729 |