Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166243.2 | NP_001159715.1 | 6 | 533 | ||
NM_001320899.1 | NP_001307828.1 | 6 | 533 | |||
NM_001320900.1 | NP_001307829.1 | 7 | 627 | |||
NM_002012.3 | NP_002003.1 | 6 | 533 | |||
XM_017005880.1 | XP_016861369.1 | 6 | 531 | |||
XM_017005881.1 | XP_016861370.1 | 5 | 493 | |||
XM_017005882.1 | XP_016861371.1 | 6 | 531 | |||
XM_017005883.1 | XP_016861372.1 | 6 | 531 | |||
XM_017005884.1 | XP_016861373.1 | 6 | 531 | |||
XM_017005885.1 | XP_016861374.1 | 2 | 438 | |||
XM_017005886.1 | XP_016861375.1 | 2 | 1198 | |||
GenBank mRNA | AK289824.1 | - | 6 | 531 | ||
BC032336.1 | - | 6 | 485 | 5185162 | ||
DA239143.1 | - | 6 | 531 | |||
DQ120721.1 | - | 2 | 160 | |||
KJ534835.1 | - | 2 | 160 | |||
U46922.1 | - | 6 | 522 |