Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
siRNA ID
HSS176945
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
8
987
NM_016260.2
NP_057344.2
7
918
XM_005246384.4
XP_005246441.1
7
1028
XM_005246385.3
XP_005246442.1
7
1058
XM_005246386.4
XP_005246443.1
7
909
XM_011510802.2
XP_011509104.1
8
1100
XM_011510803.2
XP_011509105.1
8
949
XM_011510804.2
XP_011509106.1
8
872
XM_011510805.2
XP_011509107.1
8
961
XM_011510807.2
XP_011509109.1
8
1136
XM_011510808.2
XP_011509110.1
8
986
XM_011510809.2
XP_011509111.1
8
998
XM_011510810.2
XP_011509112.1
8
912
XM_011510811.2
XP_011509113.1
7
1033
XM_011510812.2
XP_011509114.1
7
894
XM_011510814.2
XP_011509116.1
7
1392
XM_011510815.2
XP_011509117.1
7
840
XM_011510816.2
XP_011509118.1
5
841
XM_011510818.2
XP_011509120.1
5
6495
XM_017003589.1
XP_016859078.1
7
980
XM_017003590.1
XP_016859079.1
7
950
XM_017003591.1
XP_016859080.1
5
1383
GenBank mRNA
AF130863.1
-
7
918
AK301503.1
-
7
914
AY008294.1
-
1
1382
AY587062.1
-
8
902
AY587063.1
-
8
864
AY587064.1
-
8
948
AY587065.1
-
7
755
AY587067.1
-
6
566
AY587068.1
-
7
826
AY587069.1
-
6
671
AY587070.1
-
4
314
AY587071.1
-
5
533
AY587073.1
-
7
928
BC028936.1
-
8
987
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0