Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136156.1 | NP_001129628.1 | 4 | 2440 | ||
NM_014910.4 | NP_055725.2 | 3 | 2346 | |||
GenBank mRNA | AB029007.1 | - | 4 | 2401 | ||
AL713775.1 | - | 4 | 2327 | |||
BC110332.1 | - | 2 | 547 | |||
BC136417.1 | - | 4 | 2376 | 9020405 | ||
BX640881.1 | - | 3 | 2308 |