Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014911.3 | NP_055726.3 | 4 | 815 | ||
GenBank mRNA | AB028971.1 | - | 4 | 694 | ||
AK056043.1 | - | 4 | 622 | |||
AK310951.1 | - | 4 | 618 | |||
BC002695.2 | - | 3 | 350 | |||
BC104842.1 | - | 3 | 334 | 8143845 | ||
BC111965.1 | - | 3 | 334 | 8327433 | ||
BC143710.1 | - | 3 | 334 | 9052224 |