Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA6 SIRNA Assays ›
Gene Symbol:
PLEKHA6
Gene Name:
pleckstrin homology domain containing A6
Gene Aliases:
PEPP-3, PEPP3
Chromosome Location:
Chr.1: 204218851 - 204378376 on Build GRCh38
UniGene ID:
Hs.253146
Species:
Human
siRNA ID
HSS176992
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014935.4
NP_055750.2
10, 11
1923
XM_005244966.4
XP_005245023.2
13, 14
2012
XM_005244967.4
XP_005245024.2
10, 11
1849
XM_005244968.4
XP_005245025.2
10, 11
1789
XM_006711212.3
XP_006711275.1
13, 14
2298
XM_006711213.3
XP_006711276.2
12, 13
2018
XM_006711214.3
XP_006711277.2
13, 14
2072
XM_006711215.3
XP_006711278.2
11, 12
1937
XM_006711217.3
XP_006711280.2
13, 14
2072
XM_006711219.3
XP_006711282.2
10, 11
1850
XM_006711221.2
XP_006711284.1
13, 14
2396
XM_006711222.3
XP_006711285.1
13, 14
2381
XM_011509295.2
XP_011507597.1
13, 14
2072
XM_011509296.2
XP_011507598.1
12, 13
1991
XM_011509297.2
XP_011507599.1
15, 16
2898
XM_011509298.2
XP_011507600.1
13, 14
2181
XM_011509299.2
XP_011507601.2
13, 14
2187
XM_011509301.2
XP_011507603.1
13, 14
6077
XM_011509304.2
XP_011507606.1
13, 14
2026
XM_017000687.1
XP_016856176.1
13, 14
2072
XM_017000688.1
XP_016856177.1
12, 13
2018
XM_017000689.1
XP_016856178.1
11, 12
1877
XM_017000690.1
XP_016856179.1
13, 14
6017
XM_017000691.1
XP_016856180.1
8, 9
4470
XM_017000692.1
XP_016856181.1
8, 9
4410
GenBank mRNA
AB023186.1
-
10, 11
1910
BC152475.1
-
10, 11
1910
8862612
DQ925685.1
-
5, 6
418
DQ925687.1
-
3, 4
277
DQ925689.1
-
4, 5
337
DQ925690.1
-
3, 4
283
DQ925691.1
-
5, 6
414
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0