Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102664.1 | NP_001096134.1 | 3 | 328 | ||
NM_014964.4 | NP_055779.2 | 6 | 1387 | |||
NM_148921.3 | NP_683723.2 | 5 | 1216 | |||
GenBank mRNA | AB028988.1 | - | 6 | 1352 | ||
AF062084.1 | - | 3 | 768 | |||
AF062085.1 | - | 4 | 942 | |||
AK001491.1 | - | 4 | 1080 | |||
AK001996.1 | - | 3 | 935 | |||
AK024115.1 | - | 5 | 1196 | |||
AK092366.1 | - | 3 | 309 | |||
AK295652.1 | - | 3 | 289 | |||
AK307455.1 | - | 3 | 287 | |||
BC070036.1 | - | 5 | 1181 | |||
BC093972.1 | - | 3 | 841 | 7939817 | ||
BC093974.1 | - | 3 | 841 | 7939819 | ||
BC143264.1 | - | 3 | 842 | 9051766 | ||
DA220261.1 | - | 3 | 309 |