Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042540.1 | NP_001036005.1 | 4 | 346 | ||
NM_001308036.1 | NP_001294965.1 | 4 | 843 | |||
NM_007362.3 | NP_031388.2 | 4 | 505 | |||
XM_011512556.2 | XP_011510858.1 | 4 | 562 | |||
XM_011512557.2 | XP_011510859.1 | 4 | 610 | |||
XM_011512558.2 | XP_011510860.1 | 4 | 452 | |||
GenBank mRNA | AK093216.1 | - | 4 | 555 | ||
AK297506.1 | - | 4 | 808 | |||
AK308192.1 | - | 4 | 509 | |||
AK315903.1 | - | 4 | 479 | |||
AK316601.1 | - | 4 | 472 | |||
AY644767.1 | - | 4 | 429 | |||
BC001255.1 | - | 4 | 441 | 3458927 | ||
BP248419.1 | - | 4 | 505 | |||
BT006842.1 | - | 4 | 415 | |||
BX537372.1 | - | 2 | 2500 | |||
BX647994.1 | - | 4 | 490 | |||
CN421262.1 | - | 4 | 291 | |||
D59253.1 | - | 4 | 451 | |||
X84157.1 | - | 4 | 444 |