Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014974.2 | NP_055789.1 | 10 | 1278 | ||
XM_005252426.3 | XP_005252483.1 | 11 | 1437 | |||
XM_005252427.3 | XP_005252484.1 | 10 | 1318 | |||
XM_005252428.4 | XP_005252485.1 | 11 | 1488 | |||
XM_011519428.2 | XP_011517730.1 | 11 | 1419 | |||
XM_011519429.2 | XP_011517731.1 | 11 | 1417 | |||
XM_011519430.2 | XP_011517732.1 | 10 | 1294 | |||
XM_011519431.2 | XP_011517733.1 | 10 | 1294 | |||
XM_011519432.1 | XP_011517734.1 | 10 | 1719 | |||
GenBank mRNA | AB023151.2 | - | 10 | 1276 | ||
AK126063.1 | - | 11 | 1324 | |||
AK310613.1 | - | 7 | 913 | |||
BC063313.1 | - | 10 | 1278 | 30349267 |