Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIMCH1 SIRNA Assays ›
Gene Symbol:
LIMCH1
Gene Name:
LIM and calponin homology domains 1
Gene Aliases:
LIMCH1A, LMO7B
Chromosome Location:
Chr.4: 41359607 - 41700044 on Build GRCh38
UniGene ID:
Hs.335163
Species:
Human
siRNA ID
HSS177078
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIMCH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112717.2
NP_001106188.1
17
2318
NM_001112718.2
NP_001106189.1
17
2318
NM_001112719.2
NP_001106190.1
12
2103
NM_001112720.2
NP_001106191.1
11
2067
NM_001289122.1
NP_001276051.1
17
2357
NM_001289124.1
NP_001276053.1
17
2402
NM_014988.3
NP_055803.2
17
2318
XM_005248057.1
XP_005248114.1
24
3950
XM_005248058.1
XP_005248115.1
24
3950
XM_005248060.1
XP_005248117.1
24
3950
XM_005248061.1
XP_005248118.1
24
3950
XM_005248067.2
XP_005248124.1
22
4824
XM_005248072.1
XP_005248129.1
17
2318
XM_005248074.1
XP_005248131.1
16
2282
XM_005248075.1
XP_005248132.1
16
2282
XM_006713996.1
XP_006714059.1
24
3950
XM_011513642.1
XP_011511944.1
25
4025
XM_011513643.1
XP_011511945.1
25
4025
XM_011513644.1
XP_011511946.1
25
4025
XM_011513645.1
XP_011511947.1
25
4025
XM_011513646.1
XP_011511948.1
25
4025
XM_011513648.1
XP_011511950.1
24
3782
XM_011513649.1
XP_011511951.1
20
3684
XM_011513651.2
XP_011511953.1
23
4904
XM_011513653.1
XP_011511955.1
18
2393
XM_011513654.1
XP_011511956.1
17
2357
XM_011513655.1
XP_011511957.1
12
2016
XM_011513656.1
XP_011511958.1
12
1977
XM_011513657.1
XP_011511959.1
11
1941
XM_011513658.2
XP_011511960.1
16
2543
XM_017007894.1
XP_016863383.1
25
4025
XM_017007895.1
XP_016863384.1
24
3950
XM_017007896.1
XP_016863385.1
25
4025
XM_017007897.1
XP_016863386.1
24
3950
XM_017007898.1
XP_016863387.1
17
2318
XM_017007899.1
XP_016863388.1
17
2318
XM_017007900.1
XP_016863389.1
17
2318
XM_017007901.1
XP_016863390.1
17
2318
XM_017007902.1
XP_016863391.1
17
2318
XM_017007903.1
XP_016863392.1
13
2052
XM_017007904.1
XP_016863393.1
12
1977
XM_017007905.1
XP_016863394.1
12
1977
GenBank mRNA
AK027231.1
-
4
323
AK295784.1
-
10
1656
AK295836.1
-
11
1766
AK299835.1
-
11
1801
AK309846.1
-
11
2009
BC029735.1
-
2
73
CR933645.1
-
17
2319
CR936601.1
-
17
2328
CR936610.1
-
17
2403
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0