Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015044.4 | NP_055859.1 | 17 | 1835 | ||
XM_017023074.1 | XP_016878563.1 | 18 | 5524 | |||
XM_017023075.1 | XP_016878564.1 | 17 | 1838 | |||
GenBank mRNA | AB029003.1 | - | 14 | 1465 | ||
AF165531.1 | - | 17 | 1773 | |||
AF190863.1 | - | 17 | 1779 | |||
AF233522.1 | - | 17 | 1778 | |||
AF323754.1 | - | 18 | 1691 | |||
AK024619.1 | - | 17 | 1834 | |||
AK093769.1 | - | 2 | 718 | |||
BC000284.1 | - | 17 | 1817 | 3356490 |