Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005165.2 | NP_005156.1 | 6 | 701 | ||
XM_005257949.2 | XP_005258006.1 | 7 | 787 | |||
XM_011524556.1 | XP_011522858.1 | 7 | 716 | |||
GenBank mRNA | AF054987.1 | - | 6 | 612 | ||
AK293453.1 | - | 6 | 568 | |||
AK293693.1 | - | 5 | 546 | |||
AK294091.1 | - | 6 | 606 | |||
AK294157.1 | - | 6 | 594 | |||
AK296039.1 | - | 5 | 401 | |||
AK296053.1 | - | 6 | 941 | |||
AK312281.1 | - | 6 | 605 | |||
BC003613.2 | - | 6 | 647 | 2987869 | ||
BC065565.1 | - | 6 | 647 | 6155379 | ||
BC103760.1 | - | 6 | 659 | |||
BC106925.1 | - | 6 | 612 | 40032967 | ||
BC106926.1 | - | 6 | 612 | 40032968 | ||
BT007006.1 | - | 5 | 556 | |||
CR541862.1 | - | 5 | 556 | |||
CR541881.1 | - | 5 | 556 |