Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194937.1 | NP_001181866.1 | 7 | 1301 | ||
NM_001194938.1 | NP_001181867.1 | 6 | 1142 | |||
NM_015100.3 | NP_055915.2 | 7 | 1328 | |||
NM_145796.3 | NP_665739.3 | 5 | 1043 | |||
NM_207171.2 | NP_997054.1 | 5 | 855 | |||
XM_005244999.2 | XP_005245056.1 | 7 | 3996 | |||
XM_005245000.4 | XP_005245057.1 | 7 | 1887 | |||
XM_005245001.2 | XP_005245058.1 | 7 | 1112 | |||
XM_005245005.1 | XP_005245062.1 | 6 | 891 | |||
XM_005245006.4 | XP_005245063.1 | 6 | 1807 | |||
XM_011509331.2 | XP_011507633.1 | 5 | 904 | |||
XM_017000744.1 | XP_016856233.1 | 7 | 1130 | |||
XM_017000745.1 | XP_016856234.1 | 7 | 3970 | |||
XM_017000746.1 | XP_016856235.1 | 7 | 1861 | |||
XM_017000747.1 | XP_016856236.1 | 6 | 1014 | |||
XM_017000748.1 | XP_016856237.1 | 6 | 874 | |||
XM_017000749.1 | XP_016856238.1 | 6 | 1245 | |||
GenBank mRNA | AB007930.1 | - | 5 | 850 | ||
AB037911.1 | - | 6 | 883 | |||
AB075477.1 | - | 6 | 883 | |||
AJ242979.1 | - | 7 | 1319 | |||
AK000199.1 | - | 7 | 1042 | |||
AK300307.1 | - | 7 | 1015 | |||
AK302501.1 | - | 6 | 892 | |||
BC057773.1 | - | 7 | 1042 | |||
BX537561.1 | - | 5 | 757 | |||
BX537838.1 | - | 7 | 1329 | |||
BX649163.1 | - | 6 | 883 |