Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLANA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005511.1 | NP_005502.1 | 4 | 294 | ||
GenBank mRNA | AK312149.1 | - | 4 | 310 | ||
BC014423.1 | - | 4 | 319 | 4639927 | ||
CR450299.1 | - | 3 | 241 | |||
U06452.1 | - | 4 | 294 | |||
U06654.1 | - | 4 | 401 |