Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 43 | 7397 | ||
NM_001458.4 | NP_001449.3 | 44 | 7496 | |||
GenBank mRNA | AB208865.1 | - | 21 | 3138 | ||
AB371585.1 | - | 44 | 7516 | |||
AF089841.1 | - | 44 | 7336 | |||
AF146692.1 | - | 43 | 7400 | |||
AJ012737.1 | - | 43 | 7206 | |||
AK126540.1 | - | 1 | 768 |