Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134473.2 | NP_001127945.1 | 15 | 3461 | ||
NM_001278184.1 | NP_001265113.1 | 15 | 3515 | |||
NM_014615.3 | NP_055430.1 | 16 | 3734 | |||
XM_005255859.4 | XP_005255916.3 | 17 | 5876 | |||
XM_005255860.3 | XP_005255917.3 | 16 | 5769 | |||
XM_005255861.4 | XP_005255918.3 | 16 | 5657 | |||
XM_005255863.4 | XP_005255920.1 | 16 | 3626 | |||
XM_005255864.4 | XP_005255921.1 | 16 | 4358 | |||
XM_005255865.4 | XP_005255922.1 | 15 | 4139 | |||
XM_011522965.2 | XP_011521267.1 | 16 | 4125 | |||
XM_011522966.2 | XP_011521268.1 | 16 | 3592 | |||
XM_017023083.1 | XP_016878572.1 | 16 | 3884 | |||
GenBank mRNA | AK127344.1 | - | 5 | 2145 | ||
AK308612.1 | - | 7 | 1311 | |||
BC000753.2 | - | 4 | 703 | |||
BC009854.2 | - | 3 | 208 | |||
BC037556.1 | - | 15 | 3484 | 4994198 | ||
D80004.2 | - | 15 | 3399 |