Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 9 | 1576 | ||
XM_005245790.3 | XP_005245847.1 | 8 | 1711 | |||
XM_005245791.4 | XP_005245848.1 | 4 | 844 | |||
XM_017000757.1 | XP_016856246.1 | 9 | 1483 | |||
XM_017000758.1 | XP_016856247.1 | 8 | 1423 | |||
GenBank mRNA | AB020649.1 | - | 9 | 1352 | ||
BC016488.1 | - | 1 | 258 | |||
BC040441.1 | - | 9 | 1579 |