Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015192.3 | NP_056007.1 | 19 | 2352 | ||
NM_182734.2 | NP_877398.1 | 19 | 2352 | |||
XM_011529199.2 | XP_011527501.1 | 19 | 2312 | |||
XM_011529200.2 | XP_011527502.1 | 17 | 1976 | |||
XM_011529201.2 | XP_011527503.1 | 17 | 1786 | |||
XM_011529203.2 | XP_011527505.1 | 3 | 288 | |||
XM_017027752.1 | XP_016883241.1 | 19 | 2312 | |||
XM_017027753.1 | XP_016883242.1 | 19 | 1939 | |||
GenBank mRNA | AB011153.3 | - | 19 | 2352 | ||
AJ278313.1 | - | 19 | 1968 | |||
AJ278314.1 | - | 19 | 1968 | |||
AK023689.1 | - | 7 | 1117 | |||
AK127693.1 | - | 17 | 1767 | |||
AK299195.1 | - | 19 | 1987 | |||
AL137267.1 | - | 11 | 1187 | |||
AY004175.1 | - | 19 | 1950 | |||
BC069420.1 | - | 19 | 1968 | 7262405 | ||
BC117231.1 | - | 19 | 2023 | 40125782 |