Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184734.1 | NP_001171663.1 | 4 | 673 | ||
NM_001184735.1 | NP_001171664.1 | 4 | 673 | |||
NM_014010.4 | NP_054729.3 | 18 | 3229 | |||
NM_198186.3 | NP_937829.3 | 4 | 994 | |||
NM_198187.3 | NP_937830.3 | 4 | 673 | |||
NM_198188.2 | NP_937831.1 | 4 | 673 | |||
GenBank mRNA | AB014534.1 | - | 18 | 3229 | ||
AF116574.1 | - | 16 | 2450 | |||
AK024064.1 | - | 4 | 644 | |||
AK127350.1 | - | 17 | 2450 | |||
AK315025.1 | - | 16 | 2293 | |||
BC018759.2 | - | 12 | 1678 | |||
BC029272.1 | - | 4 | 994 | |||
BC093835.1 | - | 4 | 507 | |||
BC101667.1 | - | 4 | 506 | |||
BC143289.1 | - | 4 | 507 | |||
BC143292.1 | - | 4 | 507 | |||
BC143293.1 | - | 4 | 507 | |||
BC146756.1 | - | 18 | 3229 | 9007192 | ||
BM552790.1 | - | 4 | 656 | |||
BX649176.1 | - | 18 | 2564 |