Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002020.4 | NP_002011.2 | 27 | 3663 | ||
NM_182925.4 | NP_891555.2 | 27 | 3663 | |||
XM_011534478.2 | XP_011532780.1 | 27 | 4041 | |||
XM_011534484.2 | XP_011532786.1 | 25 | 3221 | |||
XM_017009263.1 | XP_016864752.1 | 27 | 4047 | |||
XM_017009264.1 | XP_016864753.1 | 27 | 4047 | |||
XM_017009265.1 | XP_016864754.1 | 27 | 4048 | |||
XM_017009266.1 | XP_016864755.1 | 27 | 4042 | |||
XM_017009267.1 | XP_016864756.1 | 27 | 4042 | |||
XM_017009268.1 | XP_016864757.1 | 27 | 3606 | |||
GenBank mRNA | AB209637.1 | - | 19 | 3954 | ||
AK291679.1 | - | 27 | 3639 | |||
AK309910.1 | - | 27 | 3639 | |||
AY233382.1 | - | 27 | 3604 | |||
AY233383.1 | - | 27 | 3604 | |||
U43143.1 | - | 27 | 3614 | |||
X68203.1 | - | 27 | 3603 | |||
X69878.1 | - | 27 | 3605 |