Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009185.2 | NP_001009185.1 | 14, 15 | 1534 | ||
NM_001205247.1 | NP_001192176.1 | 14, 15 | 1429 | |||
NM_001205248.1 | NP_001192177.1 | 14, 15 | 1521 | |||
NM_001205250.1 | NP_001192179.1 | 14, 15 | 1531 | |||
NM_001205251.1 | NP_001192180.1 | 13, 14 | 1296 | |||
NM_015256.3 | NP_056071.2 | 14, 15 | 1534 | |||
GenBank mRNA | AB020644.1 | - | 14, 15 | 1494 | ||
AF129166.1 | - | 14, 15 | 1464 | |||
AK122702.1 | - | 3, 4 | 992 | |||
AK294291.1 | - | 14, 15 | 1404 | |||
AK314452.1 | - | 14, 15 | 1436 | |||
BC026161.1 | - | 14, 15 | 1430 | 4830357 | ||
BC047453.1 | - | 13, 14 | 1218 | 5270879 | ||
DQ083031.1 | - | 14, 15 | 1395 |