Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015261.2 | NP_056076.1 | 34 | 4958 | ||
XM_011542719.2 | XP_011541021.1 | 33 | 4219 | |||
GenBank mRNA | AF070553.1 | - | 8 | 871 | ||
AK025549.1 | - | 8 | 988 | |||
AK025667.1 | - | 18 | 2249 | |||
AK095267.1 | - | 4 | 2101 | |||
AK124878.1 | - | 34 | 4884 | |||
BC011408.1 | - | 12 | 1533 | |||
BC098398.1 | - | 34 | 4417 | 30528692 | ||
BC144524.1 | - | 35 | 4783 | |||
D29954.1 | - | 34 | 4380 |