Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 4, 5 | 495 | ||
NM_001171945.1 | NP_001165416.1 | 6, 7 | 774 | |||
NM_001171946.1 | NP_001165417.1 | 5, 6 | 542 | |||
NM_025154.5 | NP_079430.3 | 4, 5 | 457 | |||
GenBank mRNA | AB018353.1 | - | 5, 6 | 512 | ||
AB648918.1 | - | 4, 5 | 475 | |||
AK022469.1 | - | 5, 6 | 522 | |||
AK022816.1 | - | 4, 5 | 449 | |||
AK309120.1 | - | 6, 7 | 746 | |||
BC013613.1 | - | 5, 6 | 542 | |||
BC142707.1 | - | 4, 5 | 505 | 40148817 | ||
BC152418.1 | - | 4, 5 | 512 | 8862528 | ||
DA738650.1 | - | 4, 5 | 457 |