Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015302.1 | NP_056117.1 | 2 | 189 | ||
GenBank mRNA | AB020648.1 | - | 2 | 162 | ||
AK307703.1 | - | 2 | 179 | |||
BC013947.2 | - | 2 | 161 | |||
BC064390.1 | - | 2 | 189 | |||
BC157882.1 | - | 2 | 180 | 8862704 | ||
BC171905.1 | - | 2 | 181 |