Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271872.1 | NP_001258801.1 | 7 | 1419 | ||
XM_005277497.3 | XP_005277554.1 | 7 | 1514 | |||
XM_005277499.4 | XP_005277556.1 | 4 | 413 | |||
XM_011542015.2 | XP_011540317.1 | 6 | 987 | |||
XM_017002149.1 | XP_016857638.1 | 5 | 1158 | |||
GenBank mRNA | BC112927.1 | - | 7 | 1419 |
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 6 | 1356 | ||
NM_001300952.1 | NP_001287881.1 | 6 | 1356 | |||
NM_015326.4 | NP_056141.2 | 6 | 1356 | |||
XM_005277510.2 | XP_005277567.1 | 7 | 1543 | |||
XM_005277511.2 | XP_005277568.1 | 7 | 1543 | |||
XM_005277514.1 | XP_005277571.1 | 7 | 1543 | |||
XM_005277515.1 | XP_005277572.1 | 7 | 1543 | |||
XM_011509354.1 | XP_011507656.1 | 7 | 1543 | |||
XM_011509355.1 | XP_011507657.1 | 7 | 1543 | |||
XM_011509356.2 | XP_011507658.1 | 7 | 1543 | |||
XM_011509359.1 | XP_011507661.1 | 7 | 1543 | |||
XM_017000838.1 | XP_016856327.1 | 5 | 1187 | |||
XM_017000839.1 | XP_016856328.1 | 5 | 1217 | |||
XM_017000840.1 | XP_016856329.1 | 7 | 832 | |||
XM_017000841.1 | XP_016856330.1 | 7 | 1543 | |||
GenBank mRNA | AB007925.1 | - | 6 | 886 | ||
AK293335.1 | - | 3 | 392 | |||
AK295845.1 | - | 6 | 1112 | |||
BC132872.1 | - | 6 | 884 | 40146894 | ||
BC132874.1 | - | 6 | 884 | 40146896 | ||
BC144343.1 | - | 6 | 884 | 9052863 |
Gene Symbol: SRGAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271870.2 | NP_001258799.2 | 6 | 1275 | ||
XM_005277421.3 | XP_005277478.1 | 4 | 998 | |||
XM_005277422.4 | XP_005277479.1 | 4 | 412 | |||
XM_017002085.1 | XP_016857574.1 | 6 | 1354 | |||
XM_017002086.1 | XP_016857575.1 | 6 | 1200 | |||
XM_017002087.1 | XP_016857576.1 | 6 | 1354 | |||
XM_017002088.1 | XP_016857577.1 | 7 | 1415 | |||
XM_017002089.1 | XP_016857578.1 | 8 | 1463 | |||
XM_017002090.1 | XP_016857579.1 | 7 | 1415 | |||
XM_017002091.1 | XP_016857580.1 | 8 | 1463 | |||
XM_017002092.1 | XP_016857581.1 | 4 | 998 | |||
GenBank mRNA | BQ706477.1 | - | 2 | 329 | ||
BY795901.2 | - | 4 | 420 |
Gene Symbol: SRGAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120535.1 | 5 | 1129 |