Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 5 | 794 | ||
NM_012239.5 | NP_036371.1 | 5 | 940 | |||
XM_005252835.1 | XP_005252892.1 | 5 | 940 | |||
XM_011519956.1 | XP_011518258.1 | 5 | 1732 | |||
XM_011519957.1 | XP_011518259.1 | 5 | 1561 | |||
XM_017017428.1 | XP_016872917.1 | 5 | 786 | |||
XM_017017429.1 | XP_016872918.1 | 5 | 960 | |||
XM_017017430.1 | XP_016872919.1 | 5 | 954 | |||
XM_017017431.1 | XP_016872920.1 | 5 | 685 | |||
GenBank mRNA | AF083108.2 | - | 5 | 1008 | ||
AK074992.1 | - | 2 | 243 | |||
AK298572.1 | - | 5 | 689 | |||
AK299438.1 | - | 5 | 772 | |||
AK301558.1 | - | 4 | 740 | |||
AK301979.1 | - | 5 | 928 | |||
AK316175.1 | - | 5 | 844 | |||
AL137276.1 | - | 1 | 63 | |||
AL535769.3 | - | 5 | 775 | |||
BC001042.2 | - | 5 | 923 | 2823471 | ||
BI755839.1 | - | 5 | 740 |