Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012426.4 | NP_036558.3 | 22 | 3209 | ||
GenBank mRNA | AK001297.1 | - | 11 | 1588 | ||
BC000463.1 | - | 4 | 553 | |||
BC003146.1 | - | 4 | 558 | |||
BC068974.1 | - | 22 | 3146 | 6168677 | ||
CB241776.1 | - | 2 | 461 |