Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUSC1 SIRNA Assays ›
Gene Symbol:
RUSC1
Gene Name:
RUN and SH3 domain containing 1
Gene Aliases:
NESCA
Chromosome Location:
Chr.1: 155320849 - 155331118 on Build GRCh38
UniGene ID:
Hs.226499
Species:
Human
siRNA ID
HSS177529
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUSC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105203.1
NP_001098673.1
3, 4
1695
NM_001105204.1
NP_001098674.1
3, 4
1695
NM_001105205.1
NP_001098675.1
2, 3
717
NM_001278227.1
NP_001265156.1
2, 3
760
NM_001278228.1
NP_001265157.1
2, 3
760
NM_001278229.1
NP_001265158.1
2, 3
760
NM_001278230.1
NP_001265159.1
1, 2
559
NM_014328.4
NP_055143.2
2, 3
760
NR_103478.1
1, 2
425
XM_006711254.1
XP_006711317.1
4, 5
2106
XM_006711256.1
XP_006711319.1
4, 5
2106
XM_006711257.1
XP_006711320.1
2, 3
374
XM_017000891.1
XP_016856380.1
3, 4
2276
XM_017000892.1
XP_016856381.1
2, 3
374
GenBank mRNA
AB026894.2
-
2, 3
184
AK055451.1
-
2, 3
955
AK074485.1
-
1, 2
313
AK074982.1
-
2, 3
179
AK093777.1
-
2, 3
213
AK125378.1
-
3, 4
1647
AK298730.1
-
2, 3
270
AK307736.1
-
2, 3
210
AK314559.1
-
2, 3
429
AL080083.1
-
1, 2
37
BC001045.2
-
2, 3
201
BC004293.1
-
1, 2
281
BC028055.1
-
1, 2
309
BC052277.1
-
2, 3
717
BX402179.2
-
2, 3
251
BX640612.1
-
1, 2
377
CN305634.1
-
1, 2
559
CR749671.1
-
1, 2
1266
DA131200.1
-
2, 3
270
DA751328.1
-
1, 2
391
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
positive regulation of signal transduction
actin binding
SH3/SH2 adaptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0