Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105203.1 | NP_001098673.1 | 6 | 1956 | ||
NM_001105204.1 | NP_001098674.1 | 6 | 1956 | |||
NM_001105205.1 | NP_001098675.1 | 5 | 978 | |||
NM_001278227.1 | NP_001265156.1 | 5 | 922 | |||
NM_001278228.1 | NP_001265157.1 | 4 | 1158 | |||
NM_001278229.1 | NP_001265158.1 | 5 | 1021 | |||
NM_001278230.1 | NP_001265159.1 | 4 | 820 | |||
NM_014328.4 | NP_055143.2 | 5 | 1021 | |||
NR_103478.1 | 4 | 686 | ||||
XM_006711254.1 | XP_006711317.1 | 7 | 2367 | |||
XM_006711256.1 | XP_006711319.1 | 7 | 2367 | |||
XM_006711257.1 | XP_006711320.1 | 5 | 635 | |||
XM_017000891.1 | XP_016856380.1 | 6 | 2537 | |||
XM_017000892.1 | XP_016856381.1 | 5 | 635 | |||
GenBank mRNA | AB026894.2 | - | 5 | 445 | ||
AK055451.1 | - | 5 | 1216 | |||
AK074485.1 | - | 4 | 574 | |||
AK074982.1 | - | 5 | 440 | |||
AK093777.1 | - | 4 | 611 | |||
AK125378.1 | - | 6 | 1908 | |||
AK298730.1 | - | 5 | 432 | |||
AK307736.1 | - | 4 | 608 | |||
AK314559.1 | - | 5 | 690 | |||
AL080083.1 | - | 4 | 298 | |||
BC001045.2 | - | 5 | 462 | |||
BC004293.1 | - | 4 | 542 | |||
BC028055.1 | - | 4 | 570 | |||
BC052277.1 | - | 5 | 978 | |||
BX402179.2 | - | 5 | 512 | |||
BX640612.1 | - | 4 | 638 | |||
CR749671.1 | - | 4 | 1527 | |||
DA131200.1 | - | 5 | 531 |