Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018094.4 | NP_060564.2 | 1 | 1504 | ||
GenBank mRNA | AJ251548.1 | - | 1 | 1262 | ||
AK001303.1 | - | 1 | 1442 | |||
AK023155.1 | - | 1 | 1448 | |||
AK225671.1 | - | 2 | 1376 | |||
BC036077.1 | - | 1 | 1458 | 5313878 |