Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 14 | 2113 | ||
XM_011518446.2 | XP_011516748.1 | 14 | 2128 | |||
XM_011518447.1 | XP_011516749.1 | 13 | 2089 | |||
XM_011518449.1 | XP_011516751.1 | 13 | 1996 | |||
XM_011518451.1 | XP_011516753.1 | 12 | 1984 | |||
XM_011518452.1 | XP_011516754.1 | 13 | 1879 | |||
XM_017014571.1 | XP_016870060.1 | 13 | 2086 | |||
XM_017014572.1 | XP_016870061.1 | 12 | 1855 | |||
GenBank mRNA | AF153415.1 | - | 14 | 2113 | ||
AK074248.1 | - | 4 | 411 | |||
AK172815.1 | - | 8 | 858 | |||
AK299952.1 | - | 14 | 2095 | |||
AL556162.3 | - | 8 | 824 | |||
AL833960.1 | - | 12 | 1504 | |||
BC014248.2 | - | 4 | 464 |