Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000150.2 | NP_000141.1 | 3 | 1338 | ||
NM_001040701.1 | NP_001035791.1 | 2 | 1210 | |||
XM_005259526.4 | XP_005259583.1 | 4 | 1405 | |||
XM_005259527.3 | XP_005259584.1 | 2 | 1201 | |||
XM_011527868.2 | XP_011526170.1 | 4 | 1477 | |||
XM_011527869.2 | XP_011526171.1 | 4 | 1474 | |||
XM_011527870.2 | XP_011526172.1 | 5 | 1585 | |||
XM_011527872.2 | XP_011526174.1 | 5 | 1605 | |||
XM_011527873.2 | XP_011526175.1 | 4 | 1540 | |||
XM_011527874.2 | XP_011526176.1 | 4 | 1560 | |||
XM_011527875.2 | XP_011526177.1 | 5 | 1631 | |||
XM_011527876.2 | XP_011526178.1 | 4 | 1586 | |||
XM_011527878.2 | XP_011526180.1 | 4 | 1477 | |||
XM_011527879.2 | XP_011526181.1 | 3 | 1432 | |||
GenBank mRNA | AF131211.1 | - | 4 | 419 | ||
AK098073.1 | - | 2 | 264 | |||
AK225795.1 | - | 2 | 717 | |||
AK313130.1 | - | 2 | 269 | |||
BC061700.1 | - | 2 | 1200 | 4606746 | ||
DA426469.1 | - | 3 | 432 | |||
M98825.1 | - | 1 | 158 | |||
U27331.1 | - | 1 | 143 | |||
U27332.1 | - | 1 | 143 | |||
U27333.1 | - | 3 | 1296 | |||
U27334.1 | - | 3 | 619 | |||
U27335.1 | - | 2 | 1171 | |||
U27336.1 | - | 2 | 494 | |||
U27337.1 | - | 3 | 781 |