Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203301.3 | NP_976046.1 | 3 | 1227 | ||
GenBank mRNA | AK294140.1 | - | 1 | 71 | ||
BC053537.1 | - | 3 | 1143 | 5181903 | ||
BC068566.1 | - | 3 | 1199 | 5270962 | ||
BX248772.1 | - | 3 | 881 |