Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166139.2 | NP_001159611.1 | 4 | 503 | ||
NM_153686.8 | NP_710153.2 | 4 | 503 | |||
NR_136669.1 | 3 | 423 | ||||
XM_011513821.2 | XP_011512123.1 | 4 | 381 | |||
XM_011513822.2 | XP_011512124.1 | 4 | 381 | |||
XM_011513823.2 | XP_011512125.1 | 3 | 227 | |||
XM_011513825.2 | XP_011512127.1 | 4 | 381 | |||
XM_011513826.2 | XP_011512128.1 | 4 | 381 | |||
XM_017007961.1 | XP_016863450.1 | 4 | 1915 | |||
XM_017007962.1 | XP_016863451.1 | 3 | 305 | |||
XM_017007963.1 | XP_016863452.1 | 3 | 608 | |||
XM_017007964.1 | XP_016863453.1 | 4 | 332 | |||
XM_017007965.1 | XP_016863454.1 | 4 | 1814 | |||
GenBank mRNA | AK096181.1 | - | 5 | 364 | ||
AK299939.1 | - | 3 | 305 | |||
BC037322.1 | - | 4 | 386 | 5260413 | ||
BU902997.1 | - | 5 | 512 | |||
BX354708.2 | - | 4 | 676 | |||
DA408372.1 | - | 4 | 488 |