Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164277.1 | NP_001157749.1 | 6, 7 | 1542 | ||
NM_001164278.1 | NP_001157750.1 | 6, 7 | 1226 | |||
NM_001164279.1 | NP_001157751.1 | 6, 7 | 968 | |||
NM_001164280.1 | NP_001157752.1 | 4, 5 | 984 | |||
NM_001467.5 | NP_001458.1 | 5, 6 | 1038 | |||
GenBank mRNA | AF110819.1 | - | 4, 5 | 785 | ||
AF110820.1 | - | 4, 5 | 785 | |||
AF110822.1 | - | 4, 5 | 785 | |||
AF111852.1 | - | 2, 3 | 1538 | |||
AK289642.1 | - | 6, 7 | 1510 | |||
AK298395.1 | - | 6, 7 | 922 | |||
AK300646.1 | - | 6, 7 | 934 | |||
AK312628.1 | - | 6, 7 | 1208 | |||
AY423732.1 | - | 4, 5 | 805 | |||
BC002400.2 | - | 4, 5 | 986 | 2822215 | ||
BC003589.1 | - | 4, 5 | 986 | 2822215 | ||
BC014663.1 | - | 4, 5 | 993 | 4857067 | ||
BC015650.2 | - | 5, 6 | 1458 | 4660508 | ||
BC064563.1 | - | 4, 5 | 981 | 5580134 | ||
CR456733.1 | - | 4, 5 | 785 | |||
Y15409.1 | - | 4, 5 | 954 |