Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC41A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173854.5 | NP_776253.3 | 5 | 1649 | ||
XM_005245069.2 | XP_005245126.1 | 5 | 1325 | |||
GenBank mRNA | AJ514402.1 | - | 4 | 736 | ||
AK125075.1 | - | 4 | 410 | |||
AK128069.1 | - | 4 | 902 | |||
AK314732.1 | - | 4 | 807 | |||
AL831974.1 | - | 5 | 1485 | |||
AL832001.1 | - | 5 | 1485 | |||
BC136303.1 | - | 4 | 1130 | 9020290 | ||
BX641054.1 | - | 5 | 1487 |