Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173573.2 | NP_775844.2 | 5, 6 | 609 | ||
XM_011519965.2 | XP_011518267.2 | 5, 6 | 784 | |||
XM_011519967.2 | XP_011518269.2 | 5, 6 | 784 | |||
XM_017017476.1 | XP_016872965.1 | 5, 6 | 784 | |||
XM_017017477.1 | XP_016872966.1 | 5, 6 | 769 | |||
XM_017017478.1 | XP_016872967.1 | 5, 6 | 625 | |||
XM_017017479.1 | XP_016872968.1 | 5, 6 | 784 | |||
GenBank mRNA | BC039077.1 | - | 5, 6 | 609 | 5168905 | |
BX384085.2 | - | 6, 7 | 712 |