Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002043.4 | NP_002034.3 | 3, 4 | 421 | ||
XM_011535713.2 | XP_011534015.1 | 2, 3 | 1506 | |||
GenBank mRNA | BC130352.1 | - | 3, 4 | 340 | 40146383 | |
BC130354.1 | - | 3, 4 | 340 | 40146385 | ||
M86868.1 | - | 3, 4 | 367 |