Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012435.2 | NP_036567.2 | 8, 9 | 1114 | ||
XM_011527893.2 | XP_011526195.1 | 8, 9 | 1129 | |||
XM_011527894.2 | XP_011526196.1 | 8, 9 | 1129 | |||
XM_011527895.2 | XP_011526197.1 | 8, 9 | 1129 | |||
XM_011527896.2 | XP_011526198.1 | 10, 11 | 1174 | |||
XM_017026567.1 | XP_016882056.1 | 9, 10 | 1262 | |||
GenBank mRNA | AB001451.1 | - | 8, 9 | 988 | ||
AL360254.1 | - | 6, 7 | 544 | |||
CF457169.1 | - | 50 |