Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 14 | 1512 | ||
GenBank mRNA | AF151809.1 | - | 14 | 1424 | ||
AK001087.1 | - | 14 | 1451 | |||
AK091282.1 | - | 2 | 3011 | |||
AK093144.1 | - | 2 | 2394 | |||
AK097015.1 | - | 7 | 2127 | |||
AK222657.1 | - | 14 | 1386 | |||
AK297770.1 | - | 13 | 1195 | |||
AK312773.1 | - | 14 | 1347 | |||
AY189688.1 | - | 14 | 1427 | |||
BC007830.1 | - | 14 | 1416 | 3635175 | ||
BC011681.2 | - | 14 | 1472 | 3010765 | ||
BC015200.1 | - | 14 | 1362 | 4333051 | ||
CR456483.1 | - | 14 | 1324 |