Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFFO1 SIRNA Assays ›
Gene Symbol:
IFFO1
Gene Name:
intermediate filament family orphan 1
Gene Aliases:
HOM-TES-103, IFFO
Chromosome Location:
Chr.12: 6539528 - 6556102 on Build GRCh38
UniGene ID:
Hs.15243
Species:
Human
siRNA ID
HSS177903
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFFO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039670.2
NP_001034759.1
6, 7
1293
NM_001193457.1
NP_001180386.1
6, 7
1317
NM_080730.4
NP_542768.2
6, 7
1293
NR_036467.1
6, 7
1390
XM_006718975.3
XP_006719038.1
6, 7
1309
XM_006718976.3
XP_006719039.1
5, 6
1303
XM_006718977.3
XP_006719040.1
5, 6
1303
XM_006718981.3
XP_006719044.1
7, 8
978
XM_011520947.2
XP_011519249.1
6, 7
1312
XM_011520948.2
XP_011519250.1
6, 7
1309
XM_011520949.2
XP_011519251.1
5, 6
1303
XM_011520952.2
XP_011519254.1
7, 8
978
XM_017019169.1
XP_016874658.1
6, 7
1312
XM_017019170.1
XP_016874659.1
6, 7
1115
XM_017019171.1
XP_016874660.1
6, 7
1309
XM_017019172.1
XP_016874661.1
7, 8
981
XM_017019173.1
XP_016874662.1
6, 7
612
GenBank mRNA
AF124432.1
-
6, 7
954
AK000710.1
-
1, 2
526
AK095106.1
-
2, 3
871
AK298905.1
-
6, 7
526
BC001790.1
-
6, 7
1370
BC002857.2
-
5, 6
431
BC004384.1
-
5, 6
431
BC010431.1
-
6, 7
1370
BC023664.2
-
6, 7
1370
BC063004.1
-
6, 7
568
BC071170.1
-
1, 2
442
BC103817.1
-
1, 2
447
BC103818.1
-
1, 2
447
BC110387.1
-
6, 7
1283
5183989
BC113848.1
-
1, 2
447
BC114454.1
-
1, 2
447
BM922290.1
-
466
BX327856.2
-
6, 7
509
BX648701.1
-
7, 8
879
BX648743.1
-
7, 8
879
DA268043.1
-
1, 2
508
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0