Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013943.2 | NP_039234.1 | 3, 4 | 600 | ||
GenBank mRNA | AF097330.1 | - | 3, 4 | 455 | ||
AF109196.1 | - | 3, 4 | 393 | |||
AK001624.1 | - | 3, 4 | 487 | |||
AK095959.1 | - | 3, 4 | 421 | |||
AK301466.1 | - | 3, 4 | 398 | |||
AK315051.1 | - | 3, 4 | 437 | |||
AL117424.1 | - | 3, 4 | 512 | |||
BC012444.1 | - | 3, 4 | 465 | 3861372 | ||
CR533501.1 | - | 3, 4 | 315 |