Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145795.1 | NP_001139267.1 | 2 | 707 | ||
NM_001145796.1 | NP_001139268.1 | 2 | 707 | |||
NM_001145797.1 | NP_001139269.1 | 2 | 707 | |||
NM_001145812.1 | NP_001139284.1 | 2 | 806 | |||
NM_001308293.1 | NP_001295222.1 | 4 | 963 | |||
NM_015503.2 | NP_056318.2 | 1 | 2537 | |||
XM_017023114.1 | XP_016878603.1 | 1 | 1126 | |||
XM_017023115.1 | XP_016878604.1 | 3 | 833 | |||
XM_017023116.1 | XP_016878605.1 | 2 | 2191 | |||
XM_017023117.1 | XP_016878606.1 | 1 | 1126 | |||
GenBank mRNA | AB037720.1 | - | 1 | 3726 | ||
AF227967.1 | - | 2 | 563 | |||
AF227968.1 | - | 2 | 563 | |||
AF227969.1 | - | 2 | 563 | |||
AK055104.1 | - | 4 | 874 | |||
AK290332.1 | - | 2 | 806 | |||
AL713760.1 | - | 1 | 2537 | |||
BC010704.1 | - | 1 | 260 | 3866324 |